• USA Home
  • H9526 - 2β-Hydroxytestosterone

EMAIL THIS PAGE TO A FRIEND
H9526 Sigma

2β-Hydroxytestosterone

≥98%

Synonym: 4-Androstene-2β,17β-diol-3-one

Purchase

Properties

Related Categories Androgen, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Hormones,
InChI Key   ZOIPFFUVGMVQGE-LXKHFVGGSA-N
assay   ≥98%
drug control   USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?