• USA Home
  • H9634 - Derivatized Silica Gel
EMAIL THIS PAGE TO A FRIEND
H9634 Sigma
Derivatized Silica Gel
40-63 μm mean particle size, average diameter 60 Å
Purchase
Properties
Related Categories Analytical/Chromatography, Chemical Synthesis, Modified Silica Gels, Octadecyl -C18- silica, Reagents,
extent of labeling   ~12% carbon loading
mean particle size   40-63 μm
pore size   60 Å average diameter
surface area   500 m2/g
Description
Preparation Note
Prepared by the reaction of silica gel S0507 with n-octyltrichlorosilane.
After hydrolysis of the unreacted chloro groups, hydroxyl groups are blocked with trimethylsilyl groups.
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
Documents
Certificate of Analysis
Protocols & Articles
Articles
Selection Guide for Polymeric Resins in Chromatography & Purification
Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?