• USA Home
  • H9758 - His-Ser 4-methoxy-β-naphthylamide acetate salt

EMAIL THIS PAGE TO A FRIEND
H9758 Sigma

His-Ser 4-methoxy-β-naphthylamide acetate salt

  • CAS Number 100929-87-1

  • Empirical Formula (Hill Notation) C20H23N5O4

  • Molecular Weight 397.43

  •  MDL number MFCD03788129

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   APZWSSHDMOSDHZ-QJHJCNPRSA-N
storage temp.   −20°C

Description

Substrates

Substrate for dipeptidyl aminopeptidase I (cathepsin C).

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?