EMAIL THIS PAGE TO A FRIEND
H9909 Sigma

HIV-1 env antigen

≥95% (SDS-PAGE), recombinant, expressed in E. coli, Galactosidase-tagged

Synonym: gp41 HIV antigen

Purchase

Properties

Related Categories AIDS and Viral Research Reagents, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in E. coli
assay   ≥95% (SDS-PAGE)
form   solution
mol wt   mol wt 32 kDa (plus 114 kDa β-Gal tag)
concentration   ~1 mg/mL
shipped in   dry ice
storage temp.   −20°C
Gene Information   human immunodeficiency virus 1 ... env(155971)

Description

Biochem/physiol Actions

Positive control for HIV antibodies.

General description

Contains 287 amino acid residues [466-753] of HIV-1 env antigen.

Physical form

Solution in 8 M urea, 20 mM tris-HCl, pH 8.0, and 10 mM 2-mercaptoethanol.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?