EMAIL THIS PAGE TO A FRIEND
HEPII5 Sigma

Heparin−Agarose

prepacked column

Purchase

Properties

biological source   Heparin from porcine intestinal mucosa
form   prepacked column
packaging   kit of 5 columns
column volume   2.5 mL
storage temp.   2-8°C

Price and Availability

Standard component

Description

Product #

Add to Cart

Heparin−Agarose, Type II-S, saline suspension, ~170 USP units/mg ligand 5 column SDS H3025
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?