• USA Home
  • HPMALM - Hydroxypropyl methacrylate (HPMA) kit for LM

EMAIL THIS PAGE TO A FRIEND
HPMALM Sigma

Hydroxypropyl methacrylate (HPMA) kit for LM

Purchase

Properties

storage temp.   2-8°C

Description

Other Notes

Components included:
M 0908  Hydroxypropyl methacrylate (HPMA) 100 ml
E 0883  Ethylene Glycol Monobutyl ether 10 ml
T 5537  Triethylene Glycol Dimethacrylate 5 ml
P 3265  Polyethylene Glycol (PEG-400) 5 g
D 8509  N,N-Dimethylaniline 0.5 ml
B 2030  Benzoyl Peroxide 5 g

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?