• USA Home
  • HT1022 - Potassium permanganate solution

EMAIL THIS PAGE TO A FRIEND
HT1022 Sigma

Potassium permanganate solution

Purchase

Properties

InChI Key   VZJVWSHVAAUDKD-UHFFFAOYSA-N
Quality Level   GMP-IVD

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?