• USA Home
  • HT1025 - Silver nitrate solution (kit component only for kc692)

EMAIL THIS PAGE TO A FRIEND
HT1025 Sigma

Silver nitrate solution (kit component only for kc692)

Purchase

Properties

Quality Level   GMP-IVD
shipped in   wet ice
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?