• USA Home
  • HT40380 - Papanicolaou Stain, EA 50

EMAIL THIS PAGE TO A FRIEND
HT40380 Sigma

Papanicolaou Stain, EA 50

Purchase

Properties

Related Categories Cell Biology, Cytology Stains, Hematology and Histology, Routine Histology Stains
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Application

Papanicolaou stain imparts a characteristic range of coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic components.

Diagnostic Implications

For routine diagnostic cytology to aid in the identification and classification of exfoliative cells.

Other Notes

Eosin Y (certified), 0.23%,Bismarck brown (certified), 0.05%, fast green FCF (certified), 0.08%, phosphotungstic acid, 0.2%, in denatured alcohol

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol
WGK Germany 
2
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?