• USA Home
  • HT501 - Formalin Solution, Neutral Buffered

EMAIL THIS PAGE TO A FRIEND
HT501 Sigma

Formalin Solution, Neutral Buffered

Purchase

Properties

clinical   for in vitro diagnostic use
packaging   pkg of 200 10-mL vials

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

HT5011 Formalin solution, neutral buffered, 10%, case of 48 × 15 mL, histological tissue fixative

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?