• USA Home
  • HT90332 - Decolorizer Solution

EMAIL THIS PAGE TO A FRIEND
HT90332 Sigma

Decolorizer Solution

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Histology Special Stains More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label
IVD   for in vitro diagnostic use

Description

Components

Isopropyl alcohol, 75%; acetone, 25%

Application

Intended for use in the Sigma Gram Stain Procedure, HT90. This modification is based upon the work of Hucker and Conn

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
1
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?