HUTR001C Sigma

MISSION® 3′UTR Lenti GoClone

Powered by SwitchGear Genomics, Housekeeping Control

DOWNLOAD MSDS (PDF)

Synonym: HUTR

Properties

Related Categories Functional Genomics and RNAi, MISSION 3′UTR Lenti GoClone, powered by SwitchGear Genomics, Negative Controls More...
human 3′UTR sequence   ACAACGAATTTGGCTACAGCAACAGGGTGGTGGACCTCATGGCCCACATGGCCTCCAAGGAGTAAGACCCCTGGACCACCAGCCCCAGCAAGAGCACAAGAGGAAGAGAGAGACCCTCACTGCTGGGGAGTCCCTGCCACACTCAGTCCCCCACCACACTGAATCTCCCCTCCTCACAGTTGCCATGTAGACCCCTTGAAGAGGGGAGGGGCCTAGGGAGCCGCACCTTGTCATGTACCATCAATAAAGTACCCT (3′UTR sequences are truncated after the first 255 bases)
shipped in   dry ice
storage temp.   −70°C

Description

General description

Sigma® and SwitchGear Genomics have teamed up to exclusively offer a genome-wide collection of 3′UTR regions in our optimized lentivirus luciferase reporter vector system. When conducting experiments using MISSION® 3′UTR Lenti GoClones, including the proper controls permits accurate interpretation of the experimental results.

The 3′UTR of the housekeeping gene Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) was cloned into the Multiple Cloning Site (MCS) of pLSG_UTR_RenSP. This can serve as a non-targeting control for assays aimed at identifying the functional targets of a miRNA.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see issionlicense.

You may also visit http://sigmaaldrich.com/missionlicense.

GoClone is a trademark of SwitchGear Genomics

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Sigma is a registered trademark of Sigma-Aldrich Co. LLC

SwitchGear Genomics is a trademark of SwitchGear Genomics

Physical form

200 μL of at least 105 TU/mL (via p24 titering assay) lentiviral particles are provided as frozen stock.

Price and Availability

Kit component only

Description

Product #

Add to Cart

MISSION® miRNA Target ID Library 200 μL    
MISSION® Target ID Amplification Primer 1 200 μL    
MISSION® Target ID Amplification Primer 2 200 μL    

Customers Also Viewed

MISSION<SUP>®</SUP> 3′UTR Lenti GoClone<SUP>™</SUP>

Powered by SwitchGear Genomics, Empty Vector Control

Fully optimized reporter system

MISSION<SUP>®</SUP> pLKO.1-puro Luciferase shRNA Control Plasmid DNA

shRNA sequence targeting luciferase

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?