HUTR003C Sigma

MISSION® 3′UTR Lenti GoClone

Powered by SwitchGear Genomics, Non-target Control 1

DOWNLOAD MSDS (PDF)

Synonym: HUTR

Properties

Related Categories Functional Genomics and RNAi, MISSION 3′UTR Lenti GoClone, powered by SwitchGear Genomics, Negative Controls More...
human 3′UTR sequence   TTTGCAAAAATTCAGTTCTCCAATAGTACATGCCGCATTTAAACTTGAACAAACAAACCCAATCTTCTGATATTTGATAATAGCGCCTTTTAAAAACCATAATCATTAGTTTTAAAGCTCTTTGGAACATGACGGATTTAAGCACAATATGGATATTTTCtTTTTTTTTTCAAATAGAGATATCCATTTGAAAAGAAGATATTAATTCAGGATAGTAAAGCAGTATCTTCAGCTACAGTTAAGAAAAACATGTAT (3′UTR sequences are truncated after the first 255 bases)
shipped in   dry ice
storage temp.   −70°C

Description

General description

All Mission 3′UTR Lenti GoClones contain 3′UTRs that are cloned within the Multiple Cloning Site (MCS) of pLSG_UTR_RenSP. Random 3′UTR control vector HUTR003C construct contains non-conserved, non-genic, and non-repetitive human genomic fragments.

Sigma® and SwitchGear Genomics have teamed up to exclusively offer a genome-wide collection of 3′UTR regions in our optimized lentivirus luciferase reporter vector system. When conducting experiments using MISSION® 3′UTR Lenti GoClones, including the proper controls permits accurate interpretation of the experimental results.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see issionlicense.

You may also visit http://sigmaaldrich.com/missionlicense.

GoClone is a trademark of SwitchGear Genomics

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Sigma is a registered trademark of Sigma-Aldrich Co. LLC

SwitchGear Genomics is a trademark of SwitchGear Genomics

Physical form

200 μL of at least 105 TU/mL (via p24 titering assay) lentiviral particles are provided as frozen stock.

Price and Availability

Customers Also Viewed

MISSION<SUP>®</SUP> 3′UTR Lenti GoClone<SUP>™</SUP>

Powered by SwitchGear Genomics, Empty Vector Control

Fully optimized reporter system

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?