• USA Home
  • I0143 - D-myo-Inositol 1,3,4,5-tetrakis(phosphate) potassium salt

EMAIL THIS PAGE TO A FRIEND
I0143 Sigma

D-myo-Inositol 1,3,4,5-tetrakis(phosphate) potassium salt

Synonym: IP4

  • CAS Number 135269-51-1

  • Empirical Formula (Hill Notation) C6H8O18P4K8

  • Molecular Weight 804.80

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Inositol Phosphates, Lipids in Cell Signaling
storage temp.   −20°C

Description

Biochem/physiol Actions

Precursor of inositol 1,3,4-trisphosphate; increases entry of calcium across the plasma membrane; proposed regulator of intracellular calcium by control of plasma membrane transport of calcium ions.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?