• USA Home
  • I0147 - In Vitro Neurochemical Techniques
EMAIL THIS PAGE TO A FRIEND
I0147 Sigma
In Vitro Neurochemical Techniques
  •  ISBN-10 0-89603-509-3
  •  ISBN-13 978-0-89603-509-6
Purchase
Properties
Related Categories Books and Software, Cell Signaling Books, Labware More...
publication info   A.A. Boulton, et al., ed., Humana Press, 1999, 416 pp., hard cover
mfr. no.   Humana
Description
General description
Techniques range from isolation of neuronal genes and their expression to analysis of receptor-ligand interactions, to characterization of consequences of receptor activation. Methods include electrophysiological techniques to explore functional properties of receptors present in membranes of excitable cells and those to isolate novel genes central to neurobiological processes. Others cover measuring changes in gene expression, rapid identification of gene polymorphisms, and identification and characterization of second messenger pathways.
Table of Contents
Characterization of Receptors by Radiolabeled Ligand Binding Techniques.
Electrophysiological Studies on Receptors In Vitro,
Subtractive Hydridization: A Method to Identify Novel Genes Induced During Neuronal Programmed Cell Death.
Messenger RNA Differential Display: An Unbiased Tool to Uncover Alterations in Gene Expression Underlying Biological Processes,
Localization of mRNAs in Brain Sections by in situ Hybridization Using Oligonucleotide Probes.
Multiprobe Oligonucleotide Solution Hybridization for the Determination of Relative mRNA Levels,
Competitive RT-PCR to Quantitate Steady-State mRNA Levels.
A Novel PCR-RFLP Detection Method Using an Optimized Set of Restriction Enzymes,
Western Blot Analysis,
The Phosphoinositide Signal Transduction System,
Protein Phosphorylation. Immunoprecipitation of Receptors.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?