• USA Home
  • I0255 - myo-Inositol Dehydrogenase from Enterobacter aerogenes

EMAIL THIS PAGE TO A FRIEND
I0255 Sigma

myo-Inositol Dehydrogenase from Enterobacter aerogenes

lyophilized powder, ≥10 units/mg protein

Synonym: myo-Inositol: NAD+ 2-oxidoreductase, Inositol 2-dehydrogenase

Purchase

Description

Unit Definition

One unit will convert 1.0 μmole of myo-inositol and β-NAD to scyllo-inosose and β-NADH per min at pH 9.0 at 25 °C.

Physical form

Lyophilized powder containing buffer salts and stabilizers.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?