• USA Home
  • I0891 - Introduction to Protein Structure, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
I0891 Sigma
Introduction to Protein Structure, 2nd ed.
  •  ISBN-10 0-8153-2305-0
  •  ISBN-13 978-0-8153-2305-1
Purchase
Properties
Related Categories Books and Software, Labware, Proteomics Books More...
publication info   C. Branden and J. Tooze, Garland Science, 1999, 410 pp., soft cover
mfr. no.   Garland
Description
General description
This edition gives an account of the principles of protein structure, with examples of key proteins in their biological context illustrated to illuminate the structural principles described. The first chapters introduce the general principles of protein structure. Subsequent chapters use specific examples of proteins to show how they fulfill a wide range of biological functions. It concludes with chapters on the experimental approaches to determining and predicting protein structure, as well as engineering new proteins to modify their functions.
Table of Contents
Part I Basic Structural Principles
Chapter 1 The Building Blocks
Chapter 2 Motifs of Protein Structure
Chapter 3 Alpha-Domain Structures
Chapter 4 Alpha/Beta Structures
Chapter 5 Beta Structures
Chapter 6 Folding and Flexibility
Chapter 7 DNA Structures
Part II Structure, Function and Engineering
Chapter 8 DNA Recognition in Procaryotes by Helix-Turn-Helix Motifs
Chapter 9 DNA Recognition by Eucaryotic Transcription Factors
Chapter 10 Specific Transcription Factors Belong to a Few Families
Chapter 11 An Example of Enzyme Catalysis: Serine Proteinases
Chapter 12 Membrane Proteins
Chapter 13 Signal Transduction
Chapter 14 Fibrous Proteins
Chapter 15 Recognition of Foreign Molecules by the Immune System
Chapter 16 The Structure of Spherical Viruses
Chapter 17 Prediction, Engineering and Design of Protein Structures
Chapter 18 Determination of Protein Structures
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?