• USA Home
  • I1891 - Pyrophosphatase, Inorganic from baker's yeast (S. cerevisiae)

EMAIL THIS PAGE TO A FRIEND
I1891 Sigma

Pyrophosphatase, Inorganic from baker's yeast (S. cerevisiae)

≥90% (HPLC), lyophilized powder, ≥1,000 units/mg protein (BCA)

Synonym: Inorganic Pyrophosphatase, Pyrophosphate phosphohydrolase

Purchase

Description

Preparation Note

HPLC purified; Prepared from I1643

Unit Definition

One unit will liberate 1.0 μmole of inorganic orthophosphate per min at pH 7.2 at 25 °C.

Physical form

lyophilized powder containing 90% buffer salts

Application

Inorganic pyrophosphatase (PPase) is a ubiquitous enzyme catalyzing the reaction PPi + H2O → 2Pi.
It plays an important role in protein, RNA, and DNA synthesis.

General description

Molecular Weight: 71 kDa (homodimer consisting of two equal subunits of molecular weight 32-35 kDa)
Extinction Coefficient: E1% = 14.5 (280 nm)
pI: 4.75
This ubiquitous enzyme serves to drive metabolic reactions that produce pyrophosphate, since these reactions typically have equilibrium constants near unity. The catalytic mechanism has been described in the literature. Inorganic pyrophosphatase is a metalloprotease that requires Mg2+ for maximal activity. Although the hydrolysis of inorganic pyrophosphate is specific in the presence of Mg2+, both ADP and ATP can be hydrolyzed if zinc is present.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?