• USA Home
  • I2260 - N2-Isobutyryl-3′-O-benzoyl-2′-deoxyguanosine

EMAIL THIS PAGE TO A FRIEND
I2260 Sigma

N2-Isobutyryl-3′-O-benzoyl-2′-deoxyguanosine

~98%

  • CAS Number 63660-23-1

  • Empirical Formula (Hill Notation) C21H23N5O6

  • Molecular Weight 441.44

  •  EC Number 264-386-4

  •  PubChem Substance ID 24895971

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   LIOWMNSJEHKBBW-UHFFFAOYSA-N
assay   ~98%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?