• USA Home
  • I2396 - Interferon from human leukocytes

EMAIL THIS PAGE TO A FRIEND
I2396 Sigma

Interferon from human leukocytes

buffered aqueous solution

Synonym: α-IFN-Le

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones, IFN-α,
sterility   aseptically filled
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... IFNA1(3439)

Description

Analysis Note

Potency determined by viral plaque reduction method using human epithelial cells and vesicular stomatitus virus.

Unit Definition

One unit protects 50% of the indicator cells from viral cytopathology.

Physical form

Solution in phosphate buffered saline, pH ~7.3.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?