• USA Home
  • I2775 - Interleukin-1β precursor fragment 110-123

EMAIL THIS PAGE TO A FRIEND
I2775 Sigma

Interleukin-1β precursor fragment 110-123

≥97% (HPLC)

Synonym: precursor IL-1β

  • CAS Number 143305-11-7

  • Empirical Formula (Hill Notation) C68H105N21O23

  • Molecular Weight 1584.69

  •  MDL number MFCD00171230

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Substrates

Interleukin-1β converting enzyme (ICE) substrate

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?