• USA Home
  • I2877 - L-Isoleucine: D-allo-isoleucine

EMAIL THIS PAGE TO A FRIEND
I2877 Sigma

L-Isoleucine: D-allo-isoleucine

Synonym: (2RS,3S)-2-Amino-3-methylpentanoic acid

  • Empirical Formula (Hill Notation) C6H13NO2

  • Molecular Weight 131.17

  •  MDL number MFCD00064222

  •  PubChem Substance ID 24896013

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   AGPKZVBTJJNPAG-WHFBIAKZSA-N

Description

Other Notes

Approx. 1:1 mixture of diastereomers

Linkage

Often erroneously called "DL-isoleucine." See separate listing of DL-isoleucine, which is a mixture of 4 stereoisomers.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?