• USA Home
  • I2891 - Pyrophosphatase, Inorganic from Bacillus stearothermophilus

EMAIL THIS PAGE TO A FRIEND
I2891 Sigma

Pyrophosphatase, Inorganic from Bacillus stearothermophilus

lyophilized powder, 15-25 units/mg protein (biuret)

Synonym: Inorganic Pyrophosphatase, Pyrophosphate phosphohydrolase

Purchase

Description

Unit Definition

One unit will liberate 1.0 μmole of inorganic orthophosphate per min at pH 9.0 at 50 °C.

Application

Inorganic pyrophosphatase (PPase) is a ubiquitous enzyme catalyzing the reaction PPi + H2O → 2Pi.
It plays an important role in protein, RNA, and DNA synthesis.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?