Synonym: Anti-IFN-γ
MDL number MFCD00165744
Popular Documents: Datasheet (PDF)
| Related Categories | Alphabetical Index, Antibodies, Antibodies against Cytokines/Growth Factors/Chemokines for Stem Cell Biology, Antibodies for Cell Biology, Antibodies for Hematopoietic Stem Cells, |
| species reactivity | mouse |
| application(s) | immunoprecipitation: suitable |
| indirect ELISA: suitable | |
| neutralization: suitable | |
| western blot: 0.25-0.5 μg/mL using mouse interferon γ | |
| clone | H22, monoclonal |
| antibody form | purified immunoglobulin |
| form | buffered aqueous solution |
| isotype | IgG |
| mol wt | antigen mol wt ~16 kDa |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | mouse ... Ifng(15978) |
purified recombinant mouse interferon γ.
Interferons (IFNs) are cytokines that are secreted in response to viral or bacterial infections and tumorigenesis. There are two types of IFNs, type I (IFN-α and IFN-β) and type II (IFN-γ). IFN-γ binds to specific receptor complex consisting of IFNγR1 and IFNγR2.1,2 There are many pathways that are mediated by IFN-γ binding such as JAK/STAT1, AP-1, NF-κB, STAT3 and STAT5.3,4 IFN-γ is pleotropic and performs various functions related to immune response, inflammation, differentiation and activation of T cells, cell cycle and apoptosis. The most studied function of IFN-γ is priming the antigen presenting cells by upregulating major histocompatibility (MHC) Class I molecules.1,4 IFN-γ has clinical applications in autoimmune diseases (rheumatoid arthritis), multiple sclerosis, cancer, HIV and fungal infections1,5
Anti-Interferon-γ recognizes amino acid residues 15-99 of mouse Interferon-γ.
Solution, 0.2 μm filtered, in 0.01 M phosphate buffered saline, pH 7.4.
The antibody recognizes an epitope between amino acid residues 15 and 99 of mouse interferon γ.
Anti-interferon-γ antibody may be used for immunoblotting at a working concentration of 0.25-0.5 μg/ml. The antibody is suitable for neutralization reactions, ELISA and immunoprecipitation.
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?