EMAIL THIS PAGE TO A FRIEND
I3268 Sigma

Interleukin-6 human

IL-6, from human (osteosarcoma cells), solution, suitable for cell culture

Synonym: IL-6, hIL-6

Purchase

Description

Biochem/physiol Actions

Interleukin-6 (IL-6) is a multifunctional protein originally discovered in the media of cells stimulated with double stranded RNA. IL-6 appears to be directly involved in the responses that occur after infection and injury and may prove to be as important as IL-1 and TNF-α in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages, and endothelial cells. It acts upon a variety of cells, including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. IL-6 induces multiple effects, as indicated by its numerous synonyms: plasmacytoma growth factor (PCT-GF), interferon-β-2 (IFN-β2), monocyte derived human B cell growth factor, B cell stimulating factor (BSF-2), hepatocyte stimulating factor (HSF), Interleukin Hybridoma/Plasmacytoma-1 (IL-HP1). In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 also potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors.

Preparation Note

Produced in MG-63 osteosarcoma cells induced with human IL-1β.

Unit Definition

One unit equals the amount of IL-6 required to induce half maximal proliferation of T-1165 cell in vitro.

Physical form

Solution in 0.2 μm filtered phosphate buffered saline containing 0.1% human serum albumin.

Price and Availability


Filter out uncertainty.
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles

Articles

Interleukin Family (IL)

The Interleukins comprise a disparate group of cytokines and growth factors that are produced by and released from leukocytes. Interleukin-1β (1L-1β) is released primarily from stimulated macrophages...
Jennifer Fries
BioFiles 2009, 4.5, 16.
Keywords: Anti-inflammatory agents, Antitumor agents, Cancer, Cell proliferation, Gene expression, Growth factors, Transcription

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?