EMAIL THIS PAGE TO A FRIEND
I3764 Sigma

Isomaltose Agarose

Purchase

Properties

Related Categories Affinity Chromatography, Carbohydrate Matrices, Molecular Biology, Protein Chromatography, Proteomics More...
matrix   4% beaded agarose
matrix activation   epichlorohydrin
matrix spacer   3 atoms
capacity   30-70 mg/mL binding capacity (Concanavalin A)
storage temp.   −20°C

Description

Quantity

Swelling: 1 g swells to approx. 4 ml

Physical form

Lyophilized powder stabilized with lactose

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?