• USA Home
  • I4143 - Isopentenyl pyrophosphate-[1-14C] triammonium salt

EMAIL THIS PAGE TO A FRIEND
I4143 Sigma

Isopentenyl pyrophosphate-[1-14C] triammonium salt

≥95% (Radiochemical Purity, HPLC), ethanol:ammonia solution

  • CAS Number 147385-63-5

  • Empirical Formula (Hill Notation) C5H12O7P2 · 3 NH3

  • Molecular Weight 297.18

  •  MDL number MFCD00189838

  •  PubChem Substance ID 24896052

Purchase

Properties

assay   ≥95% (Radiochemical Purity, HPLC)
form   ethanol:ammonia solution
extent of labeling   40-60 mCi per mmol
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

Combi-vial

Physical form

Solution in ethanol: 0.15 M NH4OH (1:1) under N2

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?