• USA Home
  • I4500 - Inosine 5′-monophosphate disodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
I4500 Sigma

Inosine 5′-monophosphate disodium salt hydrate

from muscle, Sigma Grade, 99-100%

Synonym: 5′-IMP-Na2, 5′-Inosinic acid disodium salt hydrate, I-5′-P, IMP, Inosinic Acid

  • CAS Number 352195-40-5

  • Empirical Formula (Hill Notation) C10H11N4O8PNa2 · xH2O

  • Molecular Weight 392.17 (anhydrous basis)

  •  MDL number MFCD00150372

  •  PubChem Substance ID 24896057

Purchase

Properties

Related Categories Cell Biology, Metabolic Pathways, Metabolites and Cofactors on the Metabolic Pathways Chart, Metabolomics, Nucleotide More...
biological source   from muscle
grade   Sigma Grade
assay   99-100%
storage temp.   2-8°C

Description

Biochem/physiol Actions

Inosine monophosphate, or inosinic acid, is the ribonucleotide of hypoxanthine and the first nucleotide formed during purine synthesis.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?