• USA Home
  • I4505 - L-Isoleucine–Agarose

EMAIL THIS PAGE TO A FRIEND
I4505 Sigma

L-Isoleucine–Agarose

saline suspension

Purchase

Properties

form   saline suspension
extent of labeling   3-7 μmol per mL
matrix   4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   amino
matrix spacer   1 atom
storage temp.   2-8°C

Description

Physical form

Suspension in 2.0 M NaCl containing 0.02% thimerosal

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?