• USA Home
  • I4509 - 4,4′-isopropylidenediphenol dimethacrylate

EMAIL THIS PAGE TO A FRIEND
I4509 Sigma

4,4′-isopropylidenediphenol dimethacrylate

Synonym: Bisphenol A dimethacrylate

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

156329 Bisphenol A dimethacrylate, >98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?