EMAIL THIS PAGE TO A FRIEND
I4522 Sigma

Lead ionophore I

heptane solution

Synonym: ETH 322, ETH 332, N,N-Dioctadecyl-N′,N′-dipropyl-3,6-dioxaoctanediamide

  • CAS Number 72469-41-1

  • Empirical Formula (Hill Notation) C48H96N2O4

  • Molecular Weight 765.29

  •  Beilstein Registry Number 2494396

  •  MDL number MFCD00144579

Purchase

Properties

InChI Key   UKLVPVPGVSSSIB-UHFFFAOYSA-N
form   heptane solution
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1206 3 / PGII
WGK Germany 
3
Flash Point(F) 
24.8 °F
Flash Point(C) 
-4 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?