• USA Home
  • I4631 - IgG from equine serum

EMAIL THIS PAGE TO A FRIEND
I4631 Sigma

IgG from equine serum

reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder

Synonym: Horse IgG

Purchase

Properties

Related Categories Antibodies, Normal Human and Animal Immunoglobulins, Purified Immunoglobulins, Sera and Immunoglobulins, Supplementary Products More...
grade   reagent grade
assay   ≥95% (SDS-PAGE)
form   essentially salt-free, lyophilized powder
conjugate   unconjugated
storage temp.   2-8°C

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

I5506 IgG from bovine serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?