• USA Home
  • I4636 - Dupont Cronex Quanta III Intensifying Screens without lead blockers
EMAIL THIS PAGE TO A FRIEND
I4636 Sigma
Dupont Cronex Quanta III Intensifying Screens without lead blockers
for 5 × 7 in. and 13 × 18 cm film
Purchase
Properties
Related Categories Autoradiography, Autoradiography Intensifying Screens, Electrophoresis Supplies, Labware
availability   not available in EU
Description
General description
An ultra speed rare-earth intensifying screen for general autoradiographic use. For autoradiography exposures greater than 24 hours, we recommend the Lightning Plus intensifying screens.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?