• USA Home
  • I5018 - Anti-Interleukin-1α antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
I5018 Sigma

Anti-Interleukin-1α antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-IL-1α

Purchase

Description

Immunogen

recombinant human IL-1α.

General description

The antibody does not cross-react with human IL-1β, IL-2, IL-3, IL-6, TNF-α, TNF-β, or mouse GM-CSF.

Physical form

Solution, 0.2 μm filtered, in 0.5 mL phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Anti-Interleukin-1α antibody produced in rabbit was used in immunoblotting of anti- and pro-apoptotic proteins at a dilution of 1:1000.

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Interleukins are proinflammatory cytokines produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that are produced in response to bacterial infection or LPS. Both IL-1α and IL-1β exert their cellular effects by signaling through IL-1 receptor type 1. The effects of IL-1α are mediated by activation of NF-κB, JNK and MAPK pathways that activated the genes in response to infection and inflammation. Overproduction of IL-1α has been implicated in inflammatory disorders like rheumatoid arthritis and periodontitis.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

I8284 Anti-Interleukin-1α antibody produced in goat, affinity isolated antibody, lyophilized powder
I8534 Anti-Interleukin-1α antibody produced in goat, affinity isolated antibody, lyophilized powder
I8909 Anti-Interleukin-1α antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I9034 Anti-Interleukin-1α propeptide antibody produced in sheep, affinity isolated antibody, lyophilized powder
I7534 Monoclonal Anti-Interleukin-1α antibody produced in mouse, clone 4414, purified immunoglobulin, lyophilized powder
I7659 Monoclonal Anti-Interleukin-1α antibody produced in mouse, clone 3405, purified immunoglobulin, lyophilized powder
I7409 Monoclonal Anti-Interleukin-1α propeptide antibody produced in mouse, clone 409405, purified immunoglobulin, lyophilized powder
I3392 Anti-Interleukin-1α antibody produced in goat, IgG fraction of antiserum, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?