• USA Home
  • I5025 - L-Isoleucine-7-amido-4-methylcoumarin trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
I5025 Sigma

L-Isoleucine-7-amido-4-methylcoumarin trifluoroacetate salt

  • CAS Number 191723-53-2

  • Empirical Formula (Hill Notation) C16H20N2O3 · C2HF3O2

  • Molecular Weight 402.36

  •  MDL number MFCD00152116

  •  PubChem Substance ID 329815379

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   XPPJTDCCTDQORL-UXZWUECBSA-N
storage temp.   −20°C

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?