• USA Home
  • I5132 - 2-Iminobiotin N-hydroxysuccinimide ester hydrobromide

EMAIL THIS PAGE TO A FRIEND
I5132 Sigma

2-Iminobiotin N-hydroxysuccinimide ester hydrobromide

~95% (TLC)

Synonym: N-Hydroxysuccinimido-2-iminobiotin

  • CAS Number 76939-67-8

  • Empirical Formula (Hill Notation) C14H20N4O4S · HBr

  • Molecular Weight 421.31

  •  MDL number MFCD00078540

  •  PubChem Substance ID 329815380

Purchase

Properties

Related Categories Biotinylation, Molecular Biology, Protein Modification, Protein Structural Analysis, Proteomics,
InChI Key   AOQVRPXBTSXMRA-UYNHQEOZSA-N
assay   ~95% (TLC)
storage temp.   −20°C

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?