EMAIL THIS PAGE TO A FRIEND
I5531 Sigma

IIK7

solid

Synonym: N-Butanoyl 2-(9-methoxy-6H-iso­indolo[2,1-a]indol-11-yl)­ethan­amine

  • Empirical Formula (Hill Notation) C22H24N2O2

  • Molecular Weight 348.44

  •  MDL number MFCD03787973

  •  PubChem Substance ID 24278492

Purchase

Description

Biochem/physiol Actions

Melatonin receptor agonist

Price and Availability


Better Together

Discover your Target

Making Headway
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?