• USA Home
  • I5893 - Sigma Lite Plus Intensifying Screens without lead blockers
EMAIL THIS PAGE TO A FRIEND
I5893 Sigma
Sigma Lite Plus Intensifying Screens without lead blockers
For 5 x 7 in. and 13 x 18 cm film
Purchase
Description
General description
A rare earth intensifying screen for general autoradiographic use. An economical alternative to the Dupont Cronex Lightning Plus.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?