• USA Home
  • I6021 - Interleukin-4 Receptor Soluble Fragment human

EMAIL THIS PAGE TO A FRIEND
I6021 Sigma

Interleukin-4 Receptor Soluble Fragment human

≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder

Synonym: IL-4 sR

Purchase

Properties

Related Categories Cancer Research, Cell Biology, Cell Signaling and Neuroscience, Cytokines & Growth Factors, Cytokines and Growth Factors,
recombinant   expressed in baculovirus infected Sf21 cells
assay   ≥97% (SDS-PAGE)
form   lyophilized powder
mol wt   mol wt 30-35 kDa
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   human ... IL4R(3566)

Description

Analysis Note

The receptor mediated activity of human Interleukin 4 receptor soluble fragment is measured by its ability to inhibit the IL-4 induced proliferation of TF-1 cells.

Biochem/physiol Actions

Interleukin 4 receptor soluble fragment is a competitive inhibitor of IL-4.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 1.25 mg bovine serum albumin.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?