EMAIL THIS PAGE TO A FRIEND
I6140 Sigma
InStat
IBM version
Purchase
Properties
Related Categories Computer Software, Data Analysis software, Labware More...
Description
General description
Unlike all-inclusive programs designed for statisticians, InStat provides statistical routines for scientists, even those with limited knowledge of statistics. Little statistical jargon is used, and extensive help screens are available, eliminating the need for a manual. Data can be entered directly into the built-in spreadsheet or imported from ASCII files. Statistical tests include:
•  Single sample, unpaired, and paired t tests
•  P values and 95% confidence limits
•  Nonparametric analysis (Mann-Whitney, Wilcoxan)
•  Contingency tables: chi-square, Fisher′s exact, confidence limits for the difference between proportions
•  ANOVA, ordinary, or repeated measures of variance
•  F and p values
•  Bonferroni t tests between specified groups
•  Linear regression: slope, intercept, correlation coefficient, standard errors
•  Sample size, requirements for t test and chi-square.
System Requirements
Windows 95 or higher, 4Mb RAM, 3Mb free hard disk space.
Legal Information
InStat is a trademark of GraphPAD Software
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?