• USA Home
  • I6386 - N4-Isobutyryl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine

EMAIL THIS PAGE TO A FRIEND
I6386 Sigma

N4-Isobutyryl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine

Synonym: iBu-DMT-dC

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?