• USA Home
  • I6391 - 1-(5-Isoquinolinylsulfonyl)-3-methylpiperazine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
I6391 Sigma

1-(5-Isoquinolinylsulfonyl)-3-methylpiperazine dihydrochloride

Synonym: Iso-H-7

  • CAS Number 140663-38-3

  • Empirical Formula (Hill Notation) C14H17N3O2S · 2HCl

  • Molecular Weight 364.29

  •  Beilstein Registry Number 5466326

  •  MDL number MFCD00132899

  •  PubChem Substance ID 24278494

Purchase

Description

Biochem/physiol Actions

Less potent inhibitor of phosphokinase C than H-7

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?