• USA Home
  • I6641 - Amberlite® strongly acidic cation exchanger sodium form

EMAIL THIS PAGE TO A FRIEND
I6641 Sigma

Amberlite® strongly acidic cation exchanger sodium form

gel form, 100-200 mesh (wet)

Purchase

Properties

Related Categories Cation Exchange Media, Ion Exchange Chromatography, Molecular Biology, Protein Chromatography, Proteomics More...
form   gel form
contains   <20% moisture content
feature   effective pH range 0 ‑ 14
  operating temperature≤ 120 °C (unless otherwise indicated)
matrix   8% cross-linked polystyrene
matrix active group   sulfonic acid
particle size   100-200 mesh (wet)
capacity   1.9 meq/mL
  4.4 meq/g

Description

Application

Strongly Acidic Cation Exchanger

Other Notes

Ground and sieved Amberlite IR-120PLUS

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?