• USA Home
  • I7029 - Isoamylphosphonyl-Gly-Pro-Ala dipotassium salt

EMAIL THIS PAGE TO A FRIEND
I7029 Sigma

Isoamylphosphonyl-Gly-Pro-Ala dipotassium salt

≥95% (NMR, TLC), lyophilized powder

Purchase

Description

Amino Acid Sequence

Isoamylphosphonyl-Gly-Pro-Ala

Biochem/physiol Actions

Reported inhibitor of clostridial collagenase.

Price and Availability


Amplified Detection

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?