• USA Home
  • I7031 - Nunc-Immuno Washers

EMAIL THIS PAGE TO A FRIEND
I7031 Sigma

Nunc-Immuno Washers

12 channel washer fits the 96 well Nunc-Immuno plate or module format; adjustable to all well shapes, 1/cs

Purchase

Properties

Related Categories Antibody/Immuno Supplies and Accessories, Antibody/Immunology Supplies, Labware, Manufacturer Browser, Multiwell Plates,
sterility   non-sterile
packaging   case of 1
  pack of 1
mfr. no.   Nunc, 470175
size   96 wells
color   none
matrix active group   Non-treated surface treatment

Description

General description

• Easy to use
• Allows for a flexible washing procedure
• Uniform and efficient washing
• Very robust and reliable
• Adjustable to all well shapes
• Rack included (not autoclavable)
• No electrical connections
• Autoclavable at 121°C

Legal Information

Nunc-Immuno is a trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?