• USA Home
  • I7140 - Intrinsic factor from rat stomach

EMAIL THIS PAGE TO A FRIEND
I7140 Sigma

Intrinsic factor from rat stomach

lyophilized powder, 7,500-15,000 units/mg protein

Purchase

Properties

form   lyophilized powder
composition   Protein, 20-40% Lowry
storage temp.   −20°C
Gene Information   rat ... Gif(29319)

Description

Unit Definition

One unit will bind one nanogram of Vitamin B12 at pH 7.5 at 25°C using a modification of the procedure of Allen, R.H. and Mehlman, C.S., J. Biol. Chem., 248, 3670 (1973).

Physical form

Lyophilized powder containing potassium phosphate

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?