• USA Home
  • I7154 - 5-Iodo-3-indolyl-β -D-galactopyranoside

EMAIL THIS PAGE TO A FRIEND
I7154 Sigma

5-Iodo-3-indolyl-β -D-galactopyranoside

≥98% (HPLC), solid

Synonym: Purple-β-D-Gal

  • Empirical Formula (Hill Notation) C14H16INO6

  • Molecular Weight 421.18

  •  MDL number MFCD00798400

  •  PubChem Substance ID 24896110

Purchase

Properties

Related Categories Chromogenic, Galactoside, Glycobiology, Inhibitors and Substrates, Molecular Biology,
InChI Key   XNKCQUZRJVDUFR-MBJXGIAVSA-N
assay   ≥98% (HPLC)
form   solid
storage temp.   −20°C

Description

Application

Chromogenic substrate for β-galactosidase that produces a purple precipitate.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?