EMAIL THIS PAGE TO A FRIEND
I7750 Sigma

5-Iodoorotic acid

≥95%, crystalline

Synonym: 6-Carboxy-2,4-dihydroxy-5-iodopyrimidine

  • CAS Number 17687-22-8

  • Empirical Formula (Hill Notation) C5H3IN2O4

  • Molecular Weight 281.99

  •  MDL number MFCD00056031

Purchase

Properties

InChI Key   NWVVOUXUHGRCGE-UHFFFAOYSA-N
assay   (TLC)
  ≥95%
form   crystalline
impurities   ≤5% Orotic Acid
color   light yellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?