EMAIL THIS PAGE TO A FRIEND
I7771 Sigma

IGEPAL® CA-630

for electrophoresis

Synonym: Octylphenoxy poly(ethyleneoxy)ethanol, branched

Purchase

Description

Application

Used in denaturing (urea) IEF and two-dimensional electrophoresis.1 Helps facilitate extrusion of the IEF capillary gels from the tubes.2 Used extensively with immobilized pH gradient (IPG) gels.3 Concentrations between 2-4% (v/v) are typically used in an IEF gel.

Caution

May develop turbidity or sediment during storage. A clear liquid can be obtained on heating to 40°C.

Other Notes

Nonionic, non-denaturing detergent

Quality

Chemically indistinguishable from Nonidet P-40, which is no longer commercially available.

Legal Information

IGEPAL is a registered trademark of Rhodia Operations

Price and Availability


Ultrafree Centrifugal Filter Units
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
Flash Point(F) 
483.8 °F
Flash Point(C) 
251 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?