• USA Home
  • I7893 - Immunoassays: A Practical Approach
EMAIL THIS PAGE TO A FRIEND
I7893 Sigma
Immunoassays: A Practical Approach
  •  ISBN-10 0-19-963710-5
  •  ISBN-13 978-0-19-963710-2
Purchase
Properties
Related Categories Books and Software, Immunology Books, Labware More...
publication info   J. Gosling, ed., Oxford University Press, 2000, 304 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
This is designed to help biologists develop an immunoassay of any common format for any suitable analyte. The basic principles are described and the choices of assay types and formats listed. Methods for raising and for characterizing antibodies, for preparing radioactive, enzymatic, fluorescent and other labels, are described. Solid-phase reagents, standards and immunogens are explained, with procedures for their preparation. Most components can now be obtained commercially.
Table of Contents
Protocol List
Abbreviations
1. Analysis by specific binding,
2. Raising antibodies,
3. Choosing and characterizing antibodies,
4. Labels and endpoints,
5. Solid-phase reagents,
6. Standards and immunogens,
7. Optimization,
8. Validation,
9. Data processing,
10. Quality assurance,
A1. Suitable reagents and other requisites
A2. List of suppliers
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?